Assay Details
| Gene Symbol: SEL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244984.1 | NP_001231913.1 | 3 - 4 | 456 | 141 | |
| NM_005065.5 | NP_005056.3 | 3 - 4 | 456 | 141 | ||
| XM_005267989.3 | XP_005268046.1 | 3 - 4 | 459 | 141 | ||
| GenBank mRNA | AB020335.1 | - | 3 - 4 | 384 | 141 | |
| AF052059.1 | - | 3 - 4 | 432 | 141 | ||
| AK075511.1 | - | 3 - 4 | 435 | 141 | ||
| AY358651.1 | - | 3 - 4 | 428 | 141 | ||
| BP358857.1 | - | 3 - 4 | 456 | 141 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001372.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015566.hg.1 | ENST00000336735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015566.hg.1 | ENST00000555824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015566.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015566.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14015566.hg.1 | TC14001372.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400172682.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172682.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172682.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400172682.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400172682.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172682.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400172682.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400172682.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400172682.hg.1 | TC1400009842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000336735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000554744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000555923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | ENST00000557372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_001244984.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009842.hg.1 | NM_005065.5 | Refseq |

SEL1L
Genomic Map