Assay Details
| Gene Symbol: FOXN3-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024620.1 | 1 | 1189 | 67 | ||
| GenBank mRNA | AF118074.1 | - | 1 | 1189 | 67 | |
| BC069658.1 | - | 1 | 1134 | 67 | ||
| BC069757.1 | - | 1 | 1138 | 67 | ||
| BC069775.1 | - | 1 | 1138 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FOXN3 antisense RNA 1 | FOXN3-AS1 | -156.5kb | Sense | Chr. 14 - 89417354 - 89419793 on NCBI Build GRCh38 | |
| Downstream | EF-hand calcium binding domain 11 | EFCAB11 | C14orf143 | 217.5kb | Anti-sense | Chr. 14 - 89794984 - 89954777 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P106236 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P106236 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 42602 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 30294 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 56301 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 50982 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 50326 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 5053 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 6006 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 47829 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 51667 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 36907 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 31940 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P106236 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P106236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464291 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924850 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924844 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924845 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924849 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924846 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924805 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464301 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924853 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464249 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924790 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464275 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464292 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924840 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924805 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924853 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924846 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464291 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924845 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924790 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924844 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464275 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924840 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464292 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464301 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924850 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924849 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29924787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09464249 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P106236 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P106236 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464261 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464298 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464301 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464276 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464250 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464257 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464272 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464291 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464296 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464280 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464243 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464310 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464305 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464255 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464312 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464302 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464320 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464263 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464307 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464292 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464264 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464253 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464248 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464241 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464268 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464288 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464317 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464289 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464246 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464294 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464266 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464270 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464282 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464239 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464315 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464285 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09464286 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7976150 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7976151 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7976149 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7976148 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 220852_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 220852_at | - | - |
| Human Genome U133 Plus 2.0 Array | 220852_PM_at | - | - |
| Human Genome U133A 2.0 Array | 220852_at | - | - |
| Human Genome U133A 2.0 Array | 220852_PM_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw516205 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220852_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220852_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220852_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220852_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220852_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220852_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220852_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220852_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220852_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220852_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220852_at7 | - | - |
| Human Genome U133 Set | 220852_PM_at | - | - |
| Human Genome U133 Set | 220852_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051684 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051684 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 210489 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1764243 | NR_024620.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1764243 | AF118074.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1764243 | BC069658.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1764243 | BC069757.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1764243 | BC069775.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1784384 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1784384 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1764243 | NR_024620.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1764243 | AF118074.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1764243 | BC069658.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1764243 | BC069757.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1764243 | BC069775.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1764243 | NR_024620.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1764243 | AF118074.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1764243 | BC069658.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1764243 | BC069757.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1764243 | BC069775.1 | human_transcriptome |

FOXN3-AS2
Genomic Map