Assay Details
Gene Symbol: AGPAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018361.3 | NP_060831.2 | 4 - 5 | 802 | 111 | |
GenBank mRNA | AF375789.1 | - | 4 - 5 | 764 | 111 | |
AL136587.1 | - | 4 - 5 | 492 | 111 | ||
BC023550.2 | - | 4 - 5 | 746 | 4129973 | 111 | |
BC068519.1 | - | 4 - 5 | 466 | 111 | ||
BC080537.1 | - | 4 - 5 | 771 | 5015128 | 111 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000022.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08000353.hg.1 | TC08000022.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08000353.hg.1 | ENST00000285518.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08000353.hg.1 | ENST00000530716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08000353.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144773.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144773.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144773.hg.1 | ENST00000530716.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144773.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144773.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144773.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144773.hg.1 | ENST00000530716.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144773.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144773.hg.1 | TC0800006566.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
