Assay Details
| Gene Symbol: XDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000379.3 | NP_000370.2 | 11 - 12 | 1117 | 71 | |
| XM_011533095.2 | XP_011531397.1 | 11 - 12 | 1149 | 71 | ||
| XM_011533096.2 | XP_011531398.1 | 11 - 12 | 1148 | 71 | ||
| GenBank mRNA | D11456.2 | - | 11 - 12 | 1095 | 71 | |
| U39487.1 | - | 11 - 12 | 1072 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001714.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein phosphorylation
negative regulation of endothelial cell proliferation
purine nucleotide catabolic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
lactation
xanthine catabolic process
negative regulation of gene expression
negative regulation of endothelial cell differentiation
negative regulation of protein kinase B signaling
oxidation-reduction process
positive regulation of p38MAPK cascade
negative regulation of vascular endothelial growth factor signaling pathway
positive regulation of reactive oxygen species metabolic process
negative regulation of vasculogenesis
xanthine dehydrogenase activity
xanthine oxidase activity
iron ion binding
electron carrier activity
oxidoreductase activity, acting on CH-OH group of donors
oxidoreductase activity, acting on the aldehyde or oxo group of donors
protein homodimerization activity
molybdopterin cofactor binding
flavin adenine dinucleotide binding
2 iron, 2 sulfur cluster binding
negative regulation of protein phosphorylation
negative regulation of endothelial cell proliferation
purine nucleotide catabolic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
lactation
xanthine catabolic process
negative regulation of gene expression
negative regulation of endothelial cell differentiation
negative regulation of protein kinase B signaling
oxidation-reduction process
positive regulation of p38MAPK cascade
negative regulation of vascular endothelial growth factor signaling pathway
positive regulation of reactive oxygen species metabolic process
negative regulation of vasculogenesis
xanthine dehydrogenase activity
xanthine oxidase activity
iron ion binding
electron carrier activity
oxidoreductase activity, acting on CH-OH group of donors
oxidoreductase activity, acting on the aldehyde or oxo group of donors
protein homodimerization activity
molybdopterin cofactor binding
flavin adenine dinucleotide binding
2 iron, 2 sulfur cluster binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02027653.hg.1 | TC02001714.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027653.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027653.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000476043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000476043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000476043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027654.hg.1 | TC02001714.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027654.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02027654.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200188797.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200188797.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200188797.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200188797.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200188797.hg.1 | TC0200012163.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000476043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000476043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000476043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | ENST00000491727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012163.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200188796.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200188796.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200188796.hg.1 | ENST00000379416.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200188796.hg.1 | NM_000379.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200188796.hg.1 | TC0200012163.hg.1 | Clariom D TC |

XDH
Genomic Map