Assay Details
Gene Symbol: GPATCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018040.3 | NP_060510.1 | 6 - 7 | 1298 | 94 | |
XM_011509689.2 | XP_011507991.1 | 6 - 7 | 1508 | 94 | ||
XM_011509691.2 | XP_011507993.1 | 5 - 6 | 1325 | 94 | ||
XM_011509693.2 | XP_011507995.1 | 6 - 7 | 1508 | 94 | ||
GenBank mRNA | AK001114.1 | - | 6 - 7 | 1265 | 94 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003823.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01058462.hg.1 | TC01003823.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01058462.hg.1 | ENST00000366935.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01058462.hg.1 | ENST00000489246.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01058462.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01058467.hg.1 | TC01003823.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01058467.hg.1 | ENST00000366935.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01058467.hg.1 | ENST00000489246.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01058467.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231162.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231162.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231162.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231162.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231162.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231162.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231162.hg.1 | TC0100017321.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231169.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231169.hg.1 | ENST00000470014.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231169.hg.1 | ENST00000485274.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231169.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231169.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231169.hg.1 | ENST00000366935.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231169.hg.1 | ENST00000470014.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231169.hg.1 | ENST00000485274.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231169.hg.1 | ENST00000489246.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231169.hg.1 | NM_018040.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231169.hg.1 | TC0100017321.hg.1 | Clariom D TC |
