Assay Details
| Gene Symbol: ATP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000704.2 | NP_000695.2 | 5 - 6 | 560 | 84 | |
| GenBank mRNA | AK058032.1 | - | 5 - 6 | 560 | 84 | |
| AK223259.1 | - | 5 - 6 | 560 | 84 | ||
| AL832971.1 | - | 4 - 5 | 438 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001448.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
cellular sodium ion homeostasis
potassium ion import
establishment or maintenance of transmembrane electrochemical gradient
ATP hydrolysis coupled proton transport
cellular potassium ion homeostasis
ion transmembrane transport
sodium ion export from cell
magnesium ion binding
sodium:potassium-exchanging ATPase activity
ATP binding
hydrogen:potassium-exchanging ATPase activity
transport
cellular sodium ion homeostasis
potassium ion import
establishment or maintenance of transmembrane electrochemical gradient
ATP hydrolysis coupled proton transport
cellular potassium ion homeostasis
ion transmembrane transport
sodium ion export from cell
magnesium ion binding
sodium:potassium-exchanging ATPase activity
ATP binding
hydrogen:potassium-exchanging ATPase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000590916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000590916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000590916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000590916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000592767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | ENST00000621873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19019588.hg.1 | ENST00000262623.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19019588.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19019588.hg.1 | TC19001448.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900185782.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185782.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185782.hg.1 | TC1900010482.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900185782.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185782.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000262623.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000590916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000590916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000590916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000590916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000592767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | ENST00000621873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010482.hg.1 | NM_000704.2 | Refseq |

ATP4A
Genomic Map