Assay Details
Gene Symbol: ELAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018696.2 | NP_061166.1 | 1 - 2 | 98 | 124 | |
GenBank mRNA | AI870985.1 | - | 1 - 2 | 99 | 124 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000999.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_018696.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR18002117.hg.1 | TC18000999.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18002117.hg.1 | ENST00000269466.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18002117.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18002116.hg.1 | TC18000999.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18002116.hg.1 | ENST00000269466.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18002116.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000586966.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000586966.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000586966.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800163539.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163539.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163539.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163539.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800163539.hg.1 | TC1800009228.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800163539.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163539.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163539.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163539.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000586966.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000586966.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000586966.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009228.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800163535.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163535.hg.1 | ENST00000586966.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163535.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163535.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163535.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800163535.hg.1 | TC1800009228.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800163535.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163535.hg.1 | ENST00000586966.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163535.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163535.hg.1 | ENST00000591429.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163535.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800163532.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163532.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163532.hg.1 | NM_018696.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800163532.hg.1 | TC1800009228.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800163532.hg.1 | ENST00000269466.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163532.hg.1 | ENST00000588577.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800163532.hg.1 | NM_018696.2 | Refseq |
