Assay Details
Gene Symbol: EPHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017000534.1 | XP_016856023.1 | 11 - 12 | 2326 | 87 | |
XM_017000535.1 | XP_016856024.1 | 11 - 12 | 2286 | 87 | ||
XM_017000536.1 | XP_016856025.1 | 10 - 11 | 2134 | 87 | ||
GenBank mRNA | AK296788.1 | - | 10 - 11 | 1379 | 87 | |
BC037166.2 | - | 11 - 12 | 2174 | 5090654 | 87 | |
M59371.1 | - | 11 - 12 | 2169 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
angiogenesis
vasculogenesis
osteoblast differentiation
multicellular organism development
intrinsic apoptotic signaling pathway in response to DNA damage
regulation of lamellipodium assembly
notochord formation
viral process
cell migration
peptidyl-tyrosine phosphorylation
neural tube development
neuron differentiation
keratinocyte differentiation
osteoclast differentiation
mammary gland epithelial cell proliferation
regulation of cell adhesion mediated by integrin
post-anal tail morphogenesis
protein kinase B signaling
regulation of blood vessel endothelial cell migration
regulation of angiogenesis
bone remodeling
ephrin receptor signaling pathway
axial mesoderm formation
negative regulation of protein kinase B signaling
notochord cell development
cell chemotaxis
branching involved in mammary gland duct morphogenesis
lens fiber cell morphogenesis
regulation of ERK1 and ERK2 cascade
response to growth factor
positive regulation of establishment of protein localization to plasma membrane
activation of GTPase activity
cell-cell adhesion
transmembrane receptor protein tyrosine kinase activity
ephrin receptor activity
protein binding
ATP binding
cadherin binding involved in cell-cell adhesion
skeletal system development
angiogenesis
vasculogenesis
osteoblast differentiation
multicellular organism development
intrinsic apoptotic signaling pathway in response to DNA damage
regulation of lamellipodium assembly
notochord formation
viral process
cell migration
peptidyl-tyrosine phosphorylation
neural tube development
neuron differentiation
keratinocyte differentiation
osteoclast differentiation
mammary gland epithelial cell proliferation
regulation of cell adhesion mediated by integrin
post-anal tail morphogenesis
protein kinase B signaling
regulation of blood vessel endothelial cell migration
regulation of angiogenesis
bone remodeling
ephrin receptor signaling pathway
axial mesoderm formation
negative regulation of protein kinase B signaling
notochord cell development
cell chemotaxis
branching involved in mammary gland duct morphogenesis
lens fiber cell morphogenesis
regulation of ERK1 and ERK2 cascade
response to growth factor
positive regulation of establishment of protein localization to plasma membrane
activation of GTPase activity
cell-cell adhesion
transmembrane receptor protein tyrosine kinase activity
ephrin receptor activity
protein binding
ATP binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000461614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000461614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000461614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000462805.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000462805.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000462805.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000480202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000480202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000480202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000480202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | ENST00000480202.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000461614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000461614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000461614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000462805.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000462805.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000462805.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000480202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000480202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000480202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000480202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | ENST00000480202.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013028.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100196009.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100196009.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100196009.hg.1 | ENST00000358432.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100196009.hg.1 | NM_004431.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100196009.hg.1 | TC0100013028.hg.1 | Clariom D TC |
