Assay Details
Gene Symbol: RSPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032784.4 | NP_116173.2 | 1 - 2 | 388 | 77 | |
XM_017011378.1 | XP_016866867.1 | 1 - 2 | 620 | 77 | ||
GenBank mRNA | AF251057.1 | - | 1 - 2 | 380 | 77 | |
AI095468.1 | - | 1 - 2 | 388 | 77 | ||
AK314912.1 | - | 1 - 2 | 159 | 77 | ||
BC022367.1 | - | 2 - 3 | 498 | 4733444 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000968.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel remodeling
sprouting angiogenesis
Wnt signaling pathway
positive regulation of Wnt signaling pathway
branching involved in labyrinthine layer morphogenesis
positive regulation of canonical Wnt signaling pathway
positive regulation of non-canonical Wnt signaling pathway
positive regulation of Wnt signaling pathway, planar cell polarity pathway
receptor binding
frizzled binding
heparin binding
blood vessel remodeling
sprouting angiogenesis
Wnt signaling pathway
positive regulation of Wnt signaling pathway
branching involved in labyrinthine layer morphogenesis
positive regulation of canonical Wnt signaling pathway
positive regulation of non-canonical Wnt signaling pathway
positive regulation of Wnt signaling pathway, planar cell polarity pathway
receptor binding
frizzled binding
heparin binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000485757.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000485757.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000485757.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | ENST00000485757.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06011325.hg.1 | TC06000968.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06011325.hg.1 | ENST00000356698.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06011325.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06011328.hg.1 | TC06000968.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06011328.hg.1 | ENST00000356698.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06011328.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06011328.hg.1 | ENST00000485757.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06011328.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06011326.hg.1 | TC06000968.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06011326.hg.1 | ENST00000356698.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06011326.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06011326.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600164423.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164423.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164423.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600164423.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164423.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164423.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600164423.hg.1 | TC0600009392.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600164418.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164418.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164418.hg.1 | TC0600009392.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600164421.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164421.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600164421.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164421.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600164421.hg.1 | TC0600009392.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600164426.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164426.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164426.hg.1 | ENST00000485757.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164426.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600164426.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164426.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164426.hg.1 | ENST00000485757.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164426.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600164426.hg.1 | TC0600009392.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000485757.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000485757.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000485757.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | ENST00000485757.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009392.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600164422.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164422.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164422.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600164422.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164422.hg.1 | ENST00000368317.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164422.hg.1 | NM_032784.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600164422.hg.1 | TC0600009392.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600164420.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164420.hg.1 | ENST00000356698.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600164420.hg.1 | TC0600009392.hg.1 | Clariom D TC |
