Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011329.hg.1 |
TC06000968.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011329.hg.1 |
ENST00000356698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011329.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011329.hg.1 |
ENST00000485757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011329.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000485757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000485757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000485757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
ENST00000485757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164428.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164428.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164428.hg.1 |
ENST00000485757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164428.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164428.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164428.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164428.hg.1 |
ENST00000485757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600164428.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600164428.hg.1 |
TC0600009392.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000356698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000368317.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000485757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000485757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000485757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
ENST00000485757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009392.hg.1 |
NM_032784.4 |
Refseq |