Assay Details
| Gene Symbol: ELFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052906.4 | NP_443138.2 | 3 | 2358 | 133 | |
| GenBank mRNA | AB067491.1 | - | 1 | 1577 | 133 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ELFN2
Genomic Map