Assay Details
| Gene Symbol: MRPL47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020409.2 | NP_065142.2 | 1 - 2 | 120 | 91 | |
| GenBank mRNA | AY212270.1 | - | 1 - 2 | 97 | 91 | |
| CA428787.1 | - | 1 - 2 | 133 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002039.hg.1
Additional Information:
This assay is in the 5'UTR region of the AY212270.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03036109.hg.1 | TC03002039.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036109.hg.1 | ENST00000476781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036109.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036110.hg.1 | TC03002039.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036110.hg.1 | ENST00000259038.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036110.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036110.hg.1 | ENST00000476781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036110.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036110.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036108.hg.1 | TC03002039.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036108.hg.1 | ENST00000259038.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036108.hg.1 | ENST00000476781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036108.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198876.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198876.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198876.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198876.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198876.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198876.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198876.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198876.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198876.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198876.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198876.hg.1 | TC0300013270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300198874.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198874.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198874.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198874.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198874.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198874.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198874.hg.1 | TC0300013270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300198875.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198875.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198875.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198875.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198875.hg.1 | TC0300013270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |

MRPL47
Genomic Map