Assay Details
Gene Symbol: LINGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032808.6 | NP_116197.4 | 1 - 2 | 215 | 84 | |
GenBank mRNA | AK291363.1 | - | 1 - 2 | 181 | 84 | |
BI523562.1 | - | 1 - 2 | 246 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK291363.1, NM_032808.6 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
