Assay Details
| Gene Symbol: LEMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001552.4 | NP_001001552.3 | 2 - 3 | 224 | 102 | |
| NM_001199050.1 | NP_001185979.1 | 2 - 3 | 224 | 102 | ||
| XM_011510160.2 | XP_011508462.1 | 2 - 3 | 228 | 102 | ||
| XM_011510161.2 | XP_011508463.1 | 2 - 3 | 216 | 102 | ||
| XM_011510162.2 | XP_011508464.1 | 2 - 3 | 226 | 102 | ||
| XM_011510164.2 | XP_011508466.1 | 2 - 3 | 248 | 102 | ||
| XM_011510165.2 | XP_011508467.1 | 2 - 3 | 244 | 102 | ||
| GenBank mRNA | AB085610.1 | - | 2 - 3 | 191 | 102 | |
| AB096677.1 | - | 2 - 3 | 191 | 102 | ||
| AB096680.1 | - | 2 - 3 | 191 | 102 | ||
| AB096681.1 | - | 2 - 3 | 191 | 102 | ||
| AK026628.1 | - | 2 - 3 | 191 | 102 | ||
| AK302392.1 | - | 2 - 3 | 226 | 102 | ||
| BC036636.2 | - | 2 - 3 | 194 | 5296437 | 102 | |
| DC399934.1 | - | 2 - 3 | 226 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003747.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057484.hg.1 | TC01003747.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057484.hg.1 | ENST00000367153.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057484.hg.1 | ENST00000367154.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057484.hg.1 | ENST00000391936.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057484.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057484.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057483.hg.1 | TC01003747.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057483.hg.1 | ENST00000367151.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057483.hg.1 | ENST00000367152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057483.hg.1 | ENST00000367153.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057483.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057483.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244530.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244530.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244530.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244530.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244530.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244530.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244530.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244530.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244530.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244530.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244530.hg.1 | TC0100018555.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000391936.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | ENST00000476884.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001001552.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NM_001199052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018555.hg.1 | NR_037583.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244529.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244529.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244529.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244529.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244529.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244529.hg.1 | ENST00000367151.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244529.hg.1 | ENST00000367152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244529.hg.1 | ENST00000367153.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244529.hg.1 | NM_001199050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244529.hg.1 | NM_001199051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244529.hg.1 | TC0100018555.hg.1 | Clariom D TC |

LEMD1
Genomic Map