Assay Details
Gene Symbol: MGAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002410.4 | NP_002401.1 | 12 - 13 | 1824 | 101 | |
XM_005263668.4 | XP_005263725.1 | 13 - 14 | 2189 | 101 | ||
XM_005263669.4 | XP_005263726.1 | 13 - 14 | 2186 | 101 | ||
XM_005263670.3 | XP_005263727.1 | 13 - 14 | 1842 | 101 | ||
XM_006712534.3 | XP_006712597.1 | 14 - 15 | 2038 | 101 | ||
XM_011511199.2 | XP_011509501.1 | 14 - 15 | 2473 | 101 | ||
XM_011511200.1 | XP_011509502.1 | 13 - 14 | 1838 | 101 | ||
XM_011511201.2 | XP_011509503.1 | 13 - 14 | 1942 | 101 | ||
XM_011511202.1 | XP_011509504.1 | 13 - 14 | 1839 | 101 | ||
XM_017004147.1 | XP_016859636.1 | 14 - 15 | 2313 | 101 | ||
XM_017004148.1 | XP_016859637.1 | 13 - 14 | 2150 | 101 | ||
GenBank mRNA | D17716.1 | - | 12 - 13 | 1824 | 101 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000874.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02012924.hg.1 | TC02000874.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02012924.hg.1 | ENST00000281923.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02012924.hg.1 | ENST00000409645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02012924.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200165827.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200165827.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200165827.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200165827.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200165827.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200165827.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200165827.hg.1 | TC0200009402.hg.1 | Clariom D TC |
