Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02012919.hg.1 |
TC02000874.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02012919.hg.1 |
ENST00000281923.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02012919.hg.1 |
ENST00000409645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02012919.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000481801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000481801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000481801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000481801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000481801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000488365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000488365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
ENST00000488365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200165822.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200165822.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200165822.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200165822.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200165822.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200165822.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200165822.hg.1 |
TC0200009402.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000281923.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000409645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000468758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000481801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000481801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000481801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000481801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000481801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000488365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000488365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
ENST00000488365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009402.hg.1 |
NM_002410.4 |
Refseq |