Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007739.hg.1 |
TC03000351.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007739.hg.1 |
ENST00000288139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007739.hg.1 |
ENST00000350061.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007739.hg.1 |
ENST00000422281.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007739.hg.1 |
ENST00000544977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007739.hg.1 |
NM_000720.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007739.hg.1 |
NM_001128839.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007739.hg.1 |
NM_001128840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
ENST00000288139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
ENST00000350061.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
ENST00000422281.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
ENST00000481478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
NM_000720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
NM_001128839.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
NM_001128840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
ENST00000288139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
ENST00000350061.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
ENST00000422281.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
ENST00000481478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
NM_000720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
NM_001128839.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
NM_001128840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154393.hg.1 |
TC0300007517.hg.1 |
Clariom D TC |