Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038875.hg.1 |
TC02002468.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038875.hg.1 |
ENST00000283243.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038875.hg.1 |
ENST00000392771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038875.hg.1 |
NM_001007267.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038875.hg.1 |
NM_001195641.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038875.hg.1 |
NM_007366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227916.hg.1 |
ENST00000283243.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227916.hg.1 |
ENST00000392771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227916.hg.1 |
NM_001007267.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227916.hg.1 |
NM_001195641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227916.hg.1 |
NM_007366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227916.hg.1 |
ENST00000283243.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227916.hg.1 |
ENST00000392771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227916.hg.1 |
NM_001007267.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227916.hg.1 |
NM_001195641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227916.hg.1 |
NM_007366.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227916.hg.1 |
TC0200016742.hg.1 |
Clariom D TC |