Assay Details
| Gene Symbol: ATN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007026.1 | NP_001007027.1 | 2 - 3 | 267 | 101 | |
| NM_001940.3 | NP_001931.2 | 2 - 3 | 260 | 101 | ||
| GenBank mRNA | BC051795.2 | - | 2 - 3 | 278 | 4181592 | 101 |
| BE142829.1 | - | 2 - 3 | 286 | 101 | ||
| D31840.1 | - | 2 - 3 | 268 | 101 | ||
| D38529.1 | - | 1 - 2 | 198 | 101 | ||
| U23851.1 | - | 1 - 2 | 103 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12003288.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC051795.2,D31840.1,D38529.1,NM_001007026.1,NM_001940.3,U23851.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12001281.hg.1 | TC12003288.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001281.hg.1 | ENST00000356654.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001281.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001283.hg.1 | TC12003288.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001283.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001283.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001285.hg.1 | TC12003288.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001285.hg.1 | ENST00000356654.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001285.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001285.hg.1 | ENST00000544325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001285.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001285.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000541029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000541029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000541029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001286.hg.1 | TC12003288.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001286.hg.1 | ENST00000356654.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001286.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001286.hg.1 | ENST00000544325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001286.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001286.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146323.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146323.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146323.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146323.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146323.hg.1 | TC1200006654.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200146324.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146324.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146324.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146324.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146324.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146324.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146324.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146324.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146324.hg.1 | TC1200006654.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200146326.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146326.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146326.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146326.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146326.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146326.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146326.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146326.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146326.hg.1 | TC1200006654.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200146325.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146325.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146325.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146325.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146325.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146325.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146325.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146325.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146325.hg.1 | TC1200006654.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200146321.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146321.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146321.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146321.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146321.hg.1 | TC1200006654.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000541029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000541029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000541029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |

ATN1
Genomic Map