Assay Details
Gene Symbol: ERICH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC073916.1 | - | 1 - 2 | 193 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC073916.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
