Assay Details
Gene Symbol: ERICH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002912.4 | NP_001002912.4 | 7 - 8 | 1029 | 143 | |
XM_017000274.1 | XP_016855763.1 | 7 - 8 | 1029 | 143 | ||
XM_017000275.1 | XP_016855764.1 | 7 - 8 | 819 | 143 | ||
GenBank mRNA | AK127470.1 | - | 7 - 8 | 1029 | 143 | |
BC073916.1 | - | 2 - 3 | 397 | 143 | ||
BI832067.1 | - | 2 - 3 | 388 | 143 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002787.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01043442.hg.1 | TC01002787.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01043442.hg.1 | ENST00000326665.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043442.hg.1 | ENST00000420661.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043442.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01043440.hg.1 | TC01002787.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01043440.hg.1 | ENST00000326665.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043440.hg.1 | ENST00000420661.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043440.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01043448.hg.1 | TC01002787.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01043448.hg.1 | ENST00000326665.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043448.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01043450.hg.1 | TC01002787.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01043450.hg.1 | ENST00000326665.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043450.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01043451.hg.1 | TC01002787.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01043451.hg.1 | ENST00000326665.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043451.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01043446.hg.1 | TC01002787.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01043446.hg.1 | ENST00000326665.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043446.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01043449.hg.1 | TC01002787.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01043449.hg.1 | ENST00000326665.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043449.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01043447.hg.1 | TC01002787.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01043447.hg.1 | ENST00000326665.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043447.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209020.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209020.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209020.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209020.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209020.hg.1 | TC0100014610.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100209014.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209014.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209014.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209014.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209014.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209014.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209014.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209014.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209014.hg.1 | TC0100014610.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100209019.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209019.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209019.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209019.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209019.hg.1 | TC0100014610.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100209021.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209021.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209021.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209021.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209021.hg.1 | TC0100014610.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100209023.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209023.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209023.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209023.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209023.hg.1 | TC0100014610.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100209016.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209016.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209016.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209016.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209016.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209016.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209016.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209016.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209016.hg.1 | TC0100014610.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100209022.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209022.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209022.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209022.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209022.hg.1 | TC0100014610.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100209025.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209025.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209025.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209025.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209025.hg.1 | TC0100014610.hg.1 | Clariom D TC |
