Assay Details
Gene Symbol: ERICH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002912.4 | NP_001002912.4 | 10 - 11 | 1712 | 107 | |
XM_017000274.1 | XP_016855763.1 | 10 - 11 | 1712 | 107 | ||
XM_017000275.1 | XP_016855764.1 | 10 - 11 | 1502 | 107 | ||
GenBank mRNA | AK127470.1 | - | 10 - 11 | 1712 | 107 | |
BC073916.1 | - | 5 - 6 | 1080 | 107 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002787.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01043433.hg.1 | TC01002787.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01043433.hg.1 | ENST00000326665.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043433.hg.1 | ENST00000420661.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043433.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01043434.hg.1 | TC01002787.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01043434.hg.1 | ENST00000326665.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043434.hg.1 | ENST00000420661.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01043434.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000433746.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000479666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | ENST00000614534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014610.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209009.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209009.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209009.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209009.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209009.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209009.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209009.hg.1 | TC0100014610.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100209010.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209010.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209010.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209010.hg.1 | ENST00000326665.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209010.hg.1 | ENST00000420661.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100209010.hg.1 | NM_001002912.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100209010.hg.1 | TC0100014610.hg.1 | Clariom D TC |
