Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015783.hg.1 |
TC05001092.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015783.hg.1 |
ENST00000296824.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015783.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015779.hg.1 |
TC05001092.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015779.hg.1 |
ENST00000296824.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015779.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000441693.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000441693.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000441693.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000441693.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000565521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000565521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
ENST00000565521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009787.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006516.hg.1 |
ENST00000509920.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168988.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168988.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168988.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168988.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168988.hg.1 |
TC0500009787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000441693.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000441693.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000441693.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000441693.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000565521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000565521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
ENST00000565521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009787.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168983.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168983.hg.1 |
ENST00000441693.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168983.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168983.hg.1 |
ENST00000296824.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168983.hg.1 |
ENST00000441693.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168983.hg.1 |
NM_145265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168983.hg.1 |
TC0500009787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500006516.hg.1 |
ENST00000509920.1 |
Ensembl |