Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005837.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005837.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005837.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005837.hg.1 |
TC18000559.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
ENST00000456505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
ENST00000456505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005838.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005838.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005838.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005838.hg.1 |
TC18000559.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800161294.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161294.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161294.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161294.hg.1 |
TC1800008920.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800161294.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161294.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161294.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
ENST00000456505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
ENST00000456505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008920.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161295.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161295.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161295.hg.1 |
NR_027245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161295.hg.1 |
TC1800008920.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800161295.hg.1 |
ENST00000323355.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161295.hg.1 |
ENST00000454647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161295.hg.1 |
NR_027245.1 |
Refseq |