Assay Details
| Gene Symbol: MYH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002470.3 | NP_002461.2 | 9 - 10 | 882 | 100 | |
| XM_011523870.2 | XP_011522172.1 | 9 - 10 | 2249 | 100 | ||
| XM_011523871.2 | XP_011522173.1 | 9 - 10 | 882 | 100 | ||
| GenBank mRNA | X13988.1 | - | 8 - 9 | 878 | 100 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001135.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
protein dephosphorylation
muscle organ development
actin filament-based movement
muscle filament sliding
embryonic limb morphogenesis
sarcomere organization
ATP metabolic process
face morphogenesis
microfilament motor activity
calmodulin binding
ATP binding
myosin phosphatase activity
ATPase activity, coupled
actin filament binding
skeletal muscle contraction
protein dephosphorylation
muscle organ development
actin filament-based movement
muscle filament sliding
embryonic limb morphogenesis
sarcomere organization
ATP metabolic process
face morphogenesis
microfilament motor activity
calmodulin binding
ATP binding
myosin phosphatase activity
ATPase activity, coupled
actin filament binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17015495.hg.1 | ENST00000226209.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015495.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015495.hg.1 | TC17001135.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000577963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000577963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000577963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000577963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000577963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000579489.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000579489.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000579489.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000579489.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000579928.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000579928.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000579928.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000579928.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000579928.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000582580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000582580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000582580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000582580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176274.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176274.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176274.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176274.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176274.hg.1 | TC1700009746.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000577963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000577963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000577963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000577963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000577963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000579489.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000579489.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000579489.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000579489.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000579928.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000579928.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000579928.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000579928.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000579928.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000582580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000582580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000582580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000582580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | ENST00000583535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009746.hg.1 | NM_002470.3 | Refseq |

MYH3
Genomic Map