Assay Details
Gene Symbol: MRPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032014.2 | NP_114403.1 | 2 - 3 | 128 | ||
GenBank mRNA | AK054982.1 | - | 1 - 2 | 208 | ||
BC012167.1 | - | 2 - 3 | 4637888 | 128 | ||
BC054865.1 | - | 2 - 3 | 128 | 6456699 | 128 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07003355.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000414932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000414932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000414932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000414932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000414932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000467084.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000467084.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000467084.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000467084.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000483330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000483330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000483330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | ENST00000483330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07020512.hg.1 | TC07003355.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07020512.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07020512.hg.1 | ENST00000467084.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07020512.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000414932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000414932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000414932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000414932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000414932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000467084.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000467084.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000467084.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000467084.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000483330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000483330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000483330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | ENST00000483330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013547.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700200585.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700200585.hg.1 | ENST00000414932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700200585.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700200585.hg.1 | ENST00000467084.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700200585.hg.1 | ENST00000483330.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700200585.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700200585.hg.1 | ENST00000317534.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700200585.hg.1 | ENST00000414932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700200585.hg.1 | ENST00000418740.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700200585.hg.1 | ENST00000467084.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700200585.hg.1 | ENST00000483330.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700200585.hg.1 | NM_032014.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700200585.hg.1 | TC0700013547.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000426198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000455877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000477768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | ENST00000603700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001077664.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001204871.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013550.hg.1 | NM_001290076.1 | Refseq |
