Assay Details
Gene Symbol: MRPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032014.2 | NP_114403.1 | 3 - 4 | 273 | 130 | |
GenBank mRNA | AF161453.1 | - | 3 - 4 | 200 | 130 | |
BC012167.1 | - | 3 - 4 | 225 | 4637888 | 130 | |
BC054865.1 | - | 3 - 4 | 240 | 6456699 | 130 | |
CA413186.1 | - | 1 - 2 | 130 |
Gene Symbol: URGCP-MRPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204871.1 | NP_001191800.1 | 6 - 7 | 415 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
