Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003627.hg.1 |
ENST00000389658.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003627.hg.1 |
NM_005559.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003627.hg.1 |
TC18000309.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800154655.hg.1 |
ENST00000389658.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800154655.hg.1 |
ENST00000579014.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800154655.hg.1 |
NM_005559.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800154655.hg.1 |
TC1800007986.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800154655.hg.1 |
ENST00000389658.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800154655.hg.1 |
ENST00000579014.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800154655.hg.1 |
NM_005559.3 |
Refseq |