Assay Details
Gene Symbol: SOHLH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282147.1 | NP_001269076.1 | 1 - 2 | 134 | 92 | |
NM_017826.2 | NP_060296.2 | 1 - 2 | 134 | 92 | ||
GenBank mRNA | AK000456.1 | - | 1 - 2 | 58 | 92 | |
BC025383.2 | - | 1 - 2 | 107 | 4829809 | 92 | |
DB022954.1 | - | 1 - 2 | 135 | 92 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13001724.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK000456.1,BC025383.2,NM_001282147.1,NM_017826.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
multicellular organism development
spermatogenesis
cell differentiation
positive regulation of transcription from RNA polymerase II promoter
oogenesis
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein dimerization activity
transcription from RNA polymerase II promoter
multicellular organism development
spermatogenesis
cell differentiation
positive regulation of transcription from RNA polymerase II promoter
oogenesis
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein dimerization activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13006369.hg.1 | TC13001724.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13006369.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006369.hg.1 | ENST00000379881.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006369.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13006370.hg.1 | TC13001724.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13006370.hg.1 | ENST00000379881.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006370.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13006368.hg.1 | TC13001724.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13006368.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006368.hg.1 | ENST00000379881.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006368.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167598.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167598.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167598.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167598.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167598.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167598.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167598.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167598.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167598.hg.1 | TC1300010032.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300167599.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167599.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167599.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167599.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167599.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167599.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167599.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167599.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167599.hg.1 | TC1300010032.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300167600.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167600.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167600.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167600.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167600.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167600.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167600.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167600.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167600.hg.1 | TC1300010032.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300167601.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167601.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167601.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167601.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167601.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167601.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167601.hg.1 | TC1300010032.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
