Assay Details
Gene Symbol: SOHLH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282147.1 | NP_001269076.1 | 5 - 6 | 619 | 89 | |
NM_017826.2 | NP_060296.2 | 5 - 6 | 619 | 89 | ||
GenBank mRNA | AK000456.1 | - | 5 - 6 | 543 | 89 | |
AY884305.1 | - | 4 - 5 | 461 | 89 | ||
BC025383.2 | - | 5 - 6 | 592 | 4829809 | 89 |
Gene Symbol: CCDC169-SOHLH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198910.1 | NP_001185839.1 | 10 - 11 | 1076 | 89 | |
GenBank mRNA | AK301863.1 | - | 10 - 11 | 1063 | 89 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13001724.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
multicellular organism development
spermatogenesis
cell differentiation
positive regulation of transcription from RNA polymerase II promoter
oogenesis
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein dimerization activity
transcription from RNA polymerase II promoter
multicellular organism development
spermatogenesis
cell differentiation
positive regulation of transcription from RNA polymerase II promoter
oogenesis
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein dimerization activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13006364.hg.1 | TC13001724.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13006364.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006364.hg.1 | ENST00000379881.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006364.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167573.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167573.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167573.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167573.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167573.hg.1 | TC1300010031.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167592.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167592.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167592.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167592.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167592.hg.1 | TC1300010032.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167593.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167593.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167593.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167593.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167593.hg.1 | TC1300010032.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300167594.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167594.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167594.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167594.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167594.hg.1 | ENST00000317764.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167594.hg.1 | ENST00000379881.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300167594.hg.1 | NM_001282147.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167594.hg.1 | NM_017826.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300167594.hg.1 | TC1300010032.hg.1 | Clariom D TC |
