Assay Details
Gene Symbol: UCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003355.2 | NP_003346.2 | 4 - 5 | 722 | 86 | |
GenBank mRNA | AK025742.1 | - | 4 - 5 | 1185 | 86 | |
AK222540.1 | - | 4 - 5 | 721 | 86 | ||
AK222557.1 | - | 4 - 5 | 721 | 86 | ||
BC011737.2 | - | 4 - 5 | 685 | 4335185 | 86 | |
U76367.1 | - | 2 - 3 | 342 | 86 | ||
U82819.1 | - | 2 - 3 | 430 | 86 | ||
U94592.1 | - | 4 - 5 | 656 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002074.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to superoxide
response to hypoxia
translation
transport
mitochondrial transport
female pregnancy
aging
positive regulation of cell death
proton transport
cellular response to insulin stimulus
cellular response to amino acid starvation
negative regulation of apoptotic process
regulation of mitochondrial membrane potential
negative regulation of insulin secretion involved in cellular response to glucose stimulus
response to fatty acid
cellular response to glucose stimulus
liver regeneration
structural constituent of ribosome
protein binding
response to superoxide
response to hypoxia
translation
transport
mitochondrial transport
female pregnancy
aging
positive regulation of cell death
proton transport
cellular response to insulin stimulus
cellular response to amino acid starvation
negative regulation of apoptotic process
regulation of mitochondrial membrane potential
negative regulation of insulin secretion involved in cellular response to glucose stimulus
response to fatty acid
cellular response to glucose stimulus
liver regeneration
structural constituent of ribosome
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11024846.hg.1 | ENST00000310473.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11024846.hg.1 | ENST00000536983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11024846.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11024846.hg.1 | TC11002074.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000539330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000539330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000539330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000539764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000539764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000539764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000541027.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000541027.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000541027.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000541027.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000542615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000542615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000542615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000542615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000543714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000543714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000543714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000543714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000543714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000543714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000545212.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000545212.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000545212.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000545562.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000545562.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | ENST00000545562.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000539330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000539330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000539330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000539764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000539764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000539764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000541027.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000541027.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000541027.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000541027.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000542615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000542615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000542615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000542615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000543714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000543714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000543714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000543714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000543714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000543714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000545212.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000545212.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000545212.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000545562.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000545562.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | ENST00000545562.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011602.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100189475.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100189475.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100189475.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100189475.hg.1 | ENST00000545212.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100189475.hg.1 | ENST00000545562.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100189475.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100189475.hg.1 | ENST00000310473.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100189475.hg.1 | ENST00000536983.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100189475.hg.1 | ENST00000544615.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100189475.hg.1 | ENST00000545212.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100189475.hg.1 | ENST00000545562.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100189475.hg.1 | NM_003355.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100189475.hg.1 | TC1100011602.hg.1 | Clariom D TC |
