Assay Details
Gene Symbol: PRDM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046338.1 | 2 - 3 | 338 | 81 | ||
GenBank mRNA | AF275818.1 | - | 2 - 3 | 338 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152380.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152380.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152380.hg.1 | TC1100007408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100152382.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152382.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152382.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152382.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152382.hg.1 | TC1100007408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152386.hg.1 | TC1100007408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152381.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152381.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152381.hg.1 | TC1100007408.hg.1 | Clariom D TC |
