Assay Details
Gene Symbol: PRDM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256695.1 | NP_001243624.1 | 6 - 7 | 985 | 85 | |
NM_001256696.1 | NP_001243625.1 | 6 - 7 | 985 | 85 | ||
NR_046338.1 | 7 - 8 | 1091 | 85 | |||
XM_011520222.2 | XP_011518524.1 | 6 - 7 | 1133 | 85 | ||
XM_011520223.2 | XP_011518525.1 | 6 - 7 | 815 | 85 | ||
XM_011520224.2 | XP_011518526.1 | 6 - 7 | 1128 | 85 | ||
XM_011520225.2 | XP_011518527.1 | 6 - 7 | 765 | 85 | ||
XM_011520226.2 | XP_011518528.1 | 4 - 5 | 421 | 85 | ||
XM_011520227.2 | XP_011518529.1 | 3 - 4 | 351 | 85 | ||
XM_011520231.2 | XP_011518533.1 | 6 - 7 | 1120 | 85 | ||
GenBank mRNA | AF275818.1 | - | 7 - 8 | 1070 | 85 | |
AK094792.1 | - | 6 - 7 | 985 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152397.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152397.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152397.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152397.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152397.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152397.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152397.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152397.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152397.hg.1 | TC1100007408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100152398.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152398.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152398.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152398.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152398.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152398.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152398.hg.1 | TC1100007408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100152396.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152396.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152396.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152396.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152396.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152396.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152396.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152396.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152396.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152396.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152396.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152396.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152396.hg.1 | TC1100007408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100152400.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152400.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152400.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152400.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152400.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152400.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152400.hg.1 | TC1100007408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100152401.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152401.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152401.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152401.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152401.hg.1 | TC1100007408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
