Assay Details
| Gene Symbol: MIB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020774.3 | NP_065825.1 | 15 - 16 | 2468 | 120 | |
| XM_011526098.1 | XP_011524400.1 | 7 - 8 | 1469 | 120 | ||
| XM_017025873.1 | XP_016881362.1 | 12 - 13 | 2160 | 120 | ||
| GenBank mRNA | AB037744.1 | - | 3 - 4 | 384 | 120 | |
| AK075157.1 | - | 3 - 4 | 335 | 120 | ||
| AK091266.1 | - | 14 - 15 | 1898 | 120 | ||
| AY147849.1 | - | 15 - 16 | 2486 | 120 | ||
| AY149908.1 | - | 15 - 16 | 2213 | 120 | ||
| BC022403.1 | - | 5 - 6 | 589 | 120 | ||
| BC110581.1 | - | 15 - 16 | 2213 | 40008394 | 120 | |
| BC110582.1 | - | 15 - 16 | 2213 | 40008395 | 120 | |
| CR749635.1 | - | 12 - 13 | 1633 | 120 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK075157.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel development
in utero embryonic development
somitogenesis
neural tube formation
heart looping
endocytosis
Notch signaling pathway
protein ubiquitination
negative regulation of neuron differentiation
positive regulation of endocytosis
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
blood vessel development
in utero embryonic development
somitogenesis
neural tube formation
heart looping
endocytosis
Notch signaling pathway
protein ubiquitination
negative regulation of neuron differentiation
positive regulation of endocytosis
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000582102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000582102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146874.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146874.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146874.hg.1 | NM_020774.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146874.hg.1 | TC1800006841.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800146874.hg.1 | ENST00000261537.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146874.hg.1 | ENST00000578646.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146874.hg.1 | NM_020774.3 | Refseq |

MIB1
Genomic Map