Assay Details
Gene Symbol: MIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020774.3 | NP_065825.1 | 17 - 18 | 2840 | 83 | |
XM_011526098.1 | XP_011524400.1 | 9 - 10 | 1841 | 83 | ||
XM_017025873.1 | XP_016881362.1 | 14 - 15 | 2532 | 83 | ||
GenBank mRNA | AB037744.1 | - | 5 - 6 | 756 | 83 | |
AK075157.1 | - | 5 - 6 | 707 | 83 | ||
AK091266.1 | - | 16 - 17 | 2270 | 83 | ||
AL713705.1 | - | 2 - 3 | 269 | 83 | ||
AY147849.1 | - | 17 - 18 | 2858 | 83 | ||
AY149908.1 | - | 17 - 18 | 2585 | 83 | ||
BC022403.1 | - | 7 - 8 | 961 | 83 | ||
BC110581.1 | - | 17 - 18 | 2585 | 40008394 | 83 | |
BC110582.1 | - | 17 - 18 | 2585 | 40008395 | 83 | |
CR749635.1 | - | 14 - 15 | 2005 | 83 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000090.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel development
in utero embryonic development
somitogenesis
neural tube formation
heart looping
endocytosis
Notch signaling pathway
protein ubiquitination
negative regulation of neuron differentiation
positive regulation of endocytosis
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
blood vessel development
in utero embryonic development
somitogenesis
neural tube formation
heart looping
endocytosis
Notch signaling pathway
protein ubiquitination
negative regulation of neuron differentiation
positive regulation of endocytosis
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR18001031.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001031.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001031.hg.1 | TC18000090.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18001030.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001030.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001030.hg.1 | TC18000090.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | ENST00000582102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000577749.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578260.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | ENST00000582102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006841.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146876.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146876.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146876.hg.1 | NM_020774.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146876.hg.1 | TC1800006841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800146876.hg.1 | ENST00000261537.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146876.hg.1 | ENST00000578646.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146876.hg.1 | NM_020774.3 | Refseq |
