Assay Details
| Gene Symbol: RORC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005060.3 | NP_005051.2 | 2 - 3 | 177 | 61 | |
| XM_006711484.3 | XP_006711547.2 | 3 - 4 | 960 | 61 | ||
| GenBank mRNA | AB307696.1 | - | 2 - 3 | 69 | 61 | |
| AK223137.1 | - | 2 - 3 | 103 | 61 | ||
| AK298460.1 | - | 3 - 4 | 260 | 61 | ||
| AK315662.1 | - | 2 - 3 | 116 | 61 | ||
| BC031554.1 | - | 2 - 3 | 95 | 5186655 | 61 | |
| BC110571.2 | - | 2 - 3 | 129 | 40001043 | 61 | |
| CR457436.1 | - | 2 - 3 | 69 | 61 | ||
| U16997.1 | - | 2 - 3 | 138 | 61 | ||
More Information
Set Membership: |
|

RORC
Genomic Map