Assay Details
Gene Symbol: KLF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286818.1 | NP_001273747.1 | 1 - 2 | 187 | 145 | |
GenBank mRNA | AK302280.1 | - | 1 - 2 | 187 | 145 | |
EF208215.1 | - | 1 - 2 | 187 | 145 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK302280.1,EF208215.1,NM_001286818.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
transcription from RNA polymerase II promoter
microvillus assembly
regulation of microvillus assembly
skeletal muscle cell differentiation
positive regulation of fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
intestinal epithelial cell development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
transcription from RNA polymerase II promoter
microvillus assembly
regulation of microvillus assembly
skeletal muscle cell differentiation
positive regulation of fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
intestinal epithelial cell development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000464404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000464404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000476859.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000476859.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000477333.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000477333.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000477333.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000464404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000464404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000476859.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000476859.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000477333.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000477333.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000477333.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007491.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300150520.hg.1 | ENST00000477333.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150520.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150520.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300150520.hg.1 | ENST00000477333.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150520.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150520.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300150520.hg.1 | TC1300007491.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300150527.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150527.hg.1 | ENST00000476859.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150527.hg.1 | ENST00000477333.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150527.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150527.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300150527.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300150527.hg.1 | ENST00000377687.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150527.hg.1 | ENST00000476859.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150527.hg.1 | ENST00000477333.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150527.hg.1 | ENST00000539231.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150527.hg.1 | NM_001286818.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300150527.hg.1 | NM_001730.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300150527.hg.1 | TC1300007491.hg.1 | Clariom D TC |
