Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013255.hg.1 |
TC06004104.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013255.hg.1 |
ENST00000318981.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013255.hg.1 |
ENST00000360366.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013255.hg.1 |
ENST00000367174.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013255.hg.1 |
ENST00000456144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013255.hg.1 |
ENST00000456877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013255.hg.1 |
ENST00000528391.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013255.hg.1 |
ENST00000529824.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013255.hg.1 |
NM_012454.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
ENST00000318981.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
ENST00000360366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
ENST00000456877.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
ENST00000528391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
ENST00000528535.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
ENST00000529824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
NM_012454.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
ENST00000318981.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
ENST00000360366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
ENST00000456877.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
ENST00000528391.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
ENST00000528535.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
ENST00000529824.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
NM_012454.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199719.hg.1 |
TC0600014194.hg.1 |
Clariom D TC |