Assay Details
| Gene Symbol: ALG11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004127.2 | NP_001004127.2 | 1 - 2 | 76 | 75 | |
| GenBank mRNA | AK025456.1 | - | 1 - 2 | 49 | 75 | |
| AK296747.1 | - | 1 - 2 | 66 | 75 | ||
| BC111022.1 | - | 1 - 2 | 74 | |||
| BC142998.1 | - | 1 - 2 | 75 | 8860455 | 75 | |
| DC366525.1 | - | 1 - 2 | 76 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000220.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000616513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | ENST00000616513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002616.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002616.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002616.hg.1 | TC13000220.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002614.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002614.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002614.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002614.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002614.hg.1 | TC13000220.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300149099.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300149099.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300149099.hg.1 | ENST00000616513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300149099.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300149099.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300149099.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300149099.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300149099.hg.1 | ENST00000616513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300149099.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300149099.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300149099.hg.1 | TC1300007237.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000519151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000521776.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000523764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000616513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | ENST00000616513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NM_021645.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007237.hg.1 | NR_036571.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300149101.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300149101.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300149101.hg.1 | ENST00000521508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300149101.hg.1 | NM_001004127.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300149101.hg.1 | TC1300007237.hg.1 | Clariom D TC |

ALG11
Genomic Map