Assay Details
| Gene Symbol: CCDC28B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301011.1 | NP_001287940.1 | 4 - 5 | 873 | 91 | |
| XM_011542113.2 | XP_011540415.1 | 4 - 5 | 1632 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000461819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000461819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000461819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000461819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000461819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000461819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000469003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000469003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000469003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000469003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000483009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000483009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | ENST00000483009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013601.hg.1 | ENST00000373604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013601.hg.1 | ENST00000373604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013601.hg.1 | ENST00000421616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013601.hg.1 | ENST00000421616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013601.hg.1 | ENST00000421616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013601.hg.1 | ENST00000421616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013601.hg.1 | ENST00000421616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153855.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153855.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153855.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153855.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153855.hg.1 | TC0100007669.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100153856.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153856.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153856.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153856.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153856.hg.1 | TC0100007669.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000373602.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000421922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000461819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000461819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000461819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000461819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000461819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000461819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000469003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000469003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000469003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000469003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000483009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000483009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | ENST00000483009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_001301011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007669.hg.1 | NM_024296.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013601.hg.1 | ENST00000373604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013601.hg.1 | ENST00000373604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013601.hg.1 | ENST00000421616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013601.hg.1 | ENST00000421616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013601.hg.1 | ENST00000421616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013601.hg.1 | ENST00000421616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013601.hg.1 | ENST00000421616.1 | Ensembl |

CCDC28B
Genomic Map