Assay Details
| Gene Symbol: SCN9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002977.3 | NP_002968.1 | 23 - 24 | 4565 | 130 | |
| XM_005246757.2 | XP_005246814.1 | 23 - 24 | 4571 | 130 | ||
| XM_011511616.2 | XP_011509918.1 | 23 - 24 | 4532 | 130 | ||
| XM_011511617.2 | XP_011509919.1 | 23 - 24 | 4571 | 130 | ||
| XM_011511618.2 | XP_011509920.1 | 23 - 24 | 4538 | 130 | ||
| XM_011511619.2 | XP_011509921.1 | 23 - 24 | 4571 | 130 | ||
| XM_017004668.1 | XP_016860157.1 | 22 - 23 | 4100 | 130 | ||
| XM_017004669.1 | XP_016860158.1 | 18 - 19 | 3658 | 130 | ||
| GenBank mRNA | DQ857292.1 | - | 22 - 23 | 4265 | 130 | |
| X82835.1 | - | 22 - 23 | 4272 | 130 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002499.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sodium ion transport
inflammatory response
response to toxic substance
post-embryonic development
neuronal action potential
sensory perception of pain
regulation of ion transmembrane transport
sodium ion transmembrane transport
behavioral response to pain
regulation of postsynaptic membrane potential
membrane depolarization during action potential
voltage-gated sodium channel activity
sodium ion binding
sodium ion transport
inflammatory response
response to toxic substance
post-embryonic development
neuronal action potential
sensory perception of pain
regulation of ion transmembrane transport
sodium ion transmembrane transport
behavioral response to pain
regulation of postsynaptic membrane potential
membrane depolarization during action potential
voltage-gated sodium channel activity
sodium ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000303354.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000409672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000452182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000452182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000452182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000452182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000452182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000452182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000452182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000452182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000452182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000452182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000452182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000452182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000454569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000454569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000454569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000454569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000454569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000454569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000454569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000454569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000454569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000454569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000454569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000472119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000472119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000472119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000472119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | ENST00000472119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014819.hg.1 | NM_002977.3 | Refseq |

SCN9A
Genomic Map