Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014680.hg.1 |
TC05001011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014680.hg.1 |
ENST00000347982.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014680.hg.1 |
ENST00000354179.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014680.hg.1 |
ENST00000361032.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014680.hg.1 |
ENST00000439151.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014680.hg.1 |
NM_022455.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014680.hg.1 |
NM_172349.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500167471.hg.1 |
ENST00000347982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500167471.hg.1 |
ENST00000354179.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500167471.hg.1 |
ENST00000439151.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500167471.hg.1 |
NM_022455.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500167471.hg.1 |
NM_172349.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500167471.hg.1 |
ENST00000347982.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500167471.hg.1 |
ENST00000354179.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500167471.hg.1 |
ENST00000439151.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500167471.hg.1 |
NM_022455.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500167471.hg.1 |
NM_172349.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500167471.hg.1 |
TC0500009610.hg.1 |
Clariom D TC |