Assay Details
| Gene Symbol: RPS6KA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318361.1 | NP_001305290.1 | 1 - 2 | 141 | 89 | |
| GenBank mRNA | AK298507.1 | - | 1 - 2 | 112 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK298507.1, NM_001318361.1 transcripts.
Set Membership: |
|

RPS6KA4
Genomic Map