Assay Details
| Gene Symbol: DDX31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322341.1 | NP_001309270.1 | 1 - 2 | 538 | 149 | |
| GenBank mRNA | AK304715.1 | - | 1 - 2 | 153 | 149 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK304715.1, NM_001322341.1 transcripts.
Set Membership: |
|

DDX31
Genomic Map