Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
ENST00000338947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
ENST00000355779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
ENST00000358477.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
ENST00000379673.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
ENST00000379679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
ENST00000379681.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
ENST00000379705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
ENST00000426868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
ENST00000447043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
NM_001135955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
NM_001135956.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
NM_001135957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
NM_001135958.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
NM_003306.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
NM_016179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006651.hg.1 |
TC13000577.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000338947.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000355779.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000358477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000379673.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000379679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000379705.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000426868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000488717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000625583.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
NM_001135955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
NM_001135956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
NM_001135957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
NM_001135958.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
NM_003306.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
NM_016179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000338947.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000355779.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000358477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000379673.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000379679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000379705.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000426868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000488717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
ENST00000625583.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
NM_001135955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
NM_001135956.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
NM_001135957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
NM_001135958.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
NM_003306.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
NM_016179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158412.hg.1 |
TC1300008645.hg.1 |
Clariom D TC |