Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014811.hg.1 |
TC01000940.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014811.hg.1 |
ENST00000234677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014811.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014811.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000471705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000471705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000471705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000477544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000477544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000477544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000477544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000477544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000482384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
ENST00000482384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NR_034072.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NR_034072.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NR_034072.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009344.hg.1 |
NR_034072.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000468588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000471705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000471705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000471705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000477544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000477544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000477544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000477544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000477544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000482384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
ENST00000482384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NR_034072.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NR_034072.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NR_034072.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009344.hg.1 |
NR_034072.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166645.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166645.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166645.hg.1 |
ENST00000477544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166645.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166645.hg.1 |
ENST00000234677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166645.hg.1 |
ENST00000369923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166645.hg.1 |
ENST00000477544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166645.hg.1 |
NM_006513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166645.hg.1 |
TC0100009344.hg.1 |
Clariom D TC |