Assay Details
Gene Symbol: SARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006513.3 | NP_006504.2 | 6 - 7 | 854 | 96 | |
XM_006710813.2 | XP_006710876.1 | 6 - 7 | 854 | 96 | ||
GenBank mRNA | AK222677.1 | - | 6 - 7 | 815 | 96 | |
AK312771.1 | - | 6 - 7 | 815 | 96 | ||
BC000716.1 | - | 6 - 7 | 763 | 3505218 | 96 | |
BC009390.2 | - | 6 - 7 | 801 | 4129592 | 96 | |
BC111398.1 | - | 6 - 7 | 833 | 4133889 | 96 | |
D49914.1 | - | 6 - 7 | 757 | 96 | ||
X91257.1 | - | 6 - 7 | 829 | 96 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000940.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01014815.hg.1 | TC01000940.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01014815.hg.1 | ENST00000234677.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01014815.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01014815.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01014816.hg.1 | TC01000940.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01014816.hg.1 | ENST00000234677.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01014816.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01014816.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100166650.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166650.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166650.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166650.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166650.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100166650.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166650.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166650.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166650.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166650.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100166650.hg.1 | TC0100009344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100166649.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166649.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166649.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166649.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100166649.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166649.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166649.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100166649.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100166649.hg.1 | TC0100009344.hg.1 | Clariom D TC |
