Assay Details
Gene Symbol: SARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006710813.2 | XP_006710876.1 | 9 - 10 | 1360 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
