Assay Details
| Gene Symbol: NAA35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321881.1 | NP_001308810.1 | 9 - 10 | 815 | 101 | |
| NM_001321882.1 | NP_001308811.1 | 9 - 10 | 1033 | 101 | ||
| NM_024635.3 | NP_078911.3 | 9 - 10 | 819 | 101 | ||
| XM_005252127.3 | XP_005252184.1 | 9 - 10 | 890 | 101 | ||
| XM_017015012.1 | XP_016870501.1 | 10 - 11 | 878 | 101 | ||
| GenBank mRNA | AK056059.1 | - | 9 - 10 | 1033 | 101 | |
| AK292239.1 | - | 9 - 10 | 819 | 101 | ||
| BC117427.1 | - | 9 - 10 | 740 | 40125978 | 101 | |
| BC117429.1 | - | 9 - 10 | 736 | 40125980 | 101 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000391.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09004191.hg.1 | TC09000391.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004191.hg.1 | ENST00000361671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004191.hg.1 | ENST00000376040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004191.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152544.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900152544.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900152544.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152544.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900152544.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900152544.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152544.hg.1 | TC0900007775.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |

NAA35
Genomic Map