Assay Details
Gene Symbol: PPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002722.4 | NP_002713.1 | 1 - 2 | 45 | 155 | |
GenBank mRNA | BP394217.1 | - | 1 - 2 | 88 | 155 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001567.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_002722.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
neuropeptide signaling pathway
cell-cell signaling
digestion
feeding behavior
protein secretion
regulation of appetite
mitophagy in response to mitochondrial depolarization
xenophagy
G-protein coupled receptor binding
receptor binding
hormone activity
neuropeptide hormone activity
positive regulation of defense response to virus by host
neuropeptide signaling pathway
cell-cell signaling
digestion
feeding behavior
protein secretion
regulation of appetite
mitophagy in response to mitochondrial depolarization
xenophagy
G-protein coupled receptor binding
receptor binding
hormone activity
neuropeptide hormone activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17021045.hg.1 | ENST00000225992.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17021045.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17021045.hg.1 | TC17001567.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000587926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010806.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17021046.hg.1 | ENST00000225992.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17021046.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17021046.hg.1 | TC17001567.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700186274.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700186274.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700186274.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700186274.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700186274.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700186274.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700186274.hg.1 | TC1700010806.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700186272.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700186272.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700186272.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700186272.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700186272.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700186272.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700186272.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700186272.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700186272.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700186272.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700186272.hg.1 | TC1700010806.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000225992.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587006.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000587926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591228.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | ENST00000591703.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | NM_002722.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010806.hg.1 | NM_002722.4 | Refseq |
